Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-cyclopropyl-3-oxopropanoate 97%

Supplier: AMBEED, INC
Warning

A104862-1000G A104862-100G A104862-10G A104862-1G A104862-25G A104862-500G A104862-5G
77093-674EA 257 USD InStock
77093-674 77093-676 77093-678 77093-680 77093-682 77093-684 77093-686
Methyl-3-cyclopropyl-3-oxopropanoate 97%
Methyl-3-cyclopropyl-3-oxopropanoate
Precaución: for Research Use Only


Formula: C₇H₁₀O₃
MW: 142.1500 g/mol
MDL Number: MFCD00274090
CAS Number: 32249-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty