Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-D-glutamic acid 97%

Supplier: AMBEED, INC
Warning

A108227-10G A108227-25G A108227-500G
77093-800EA 10 USD InStock
77093-800 77093-802 77093-804 77093-806
N-(tert-Butoxycarbonyl)-D-glutamic acid 97%
N-(tert-Butoxycarbonyl)-D-glutamic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₆
MDL Number: MFCD00190790
CAS Number: 34404-28-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty