Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Amino-3-methoxyphenyl)-N,N-dimethylpiperidin-4-amine 96%

Supplier: AMBEED, INC
Warning

A108946-100MG A108946-1G A108946-250MG
77093-848EA 5 USD InStock
77093-848 77093-850 77093-852
1-(4-Amino-3-methoxyphenyl)-N,N-dimethylpiperidin-4-amine 96%
1-(4-Amino-3-methoxyphenyl)-N,N-dimethylpiperidin-4-amine
Precaución: for Research Use Only


Formula: C₁₄H₂₃N₃O
MDL Number: MFCD18995922
CAS Number: 1089279-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty