Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Allyl-p-toluenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A109089-100MG A109089-1G A109089-250MG A109089-5G
77093-862EA 8 USD InStock
77093-862 77093-864 77093-866 77093-868
N-Allyl-p-toluenesulfonamide 98%
N-Allyl-p-toluenesulfonamide
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂S
MDL Number: MFCD00182633
CAS Number: 50487-71-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty