Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycylglycine methyl ester hydrochloride 97%

Supplier: AMBEED, INC
Warning

A109280-100G A109280-10G A109280-1G A109280-250MG A109280-25G A109280-5G
77093-870EA 159 USD InStock
77093-870 77093-872 77093-874 77093-876 77093-878 77093-880
Glycylglycine methyl ester hydrochloride 97%
Glycylglycine methyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₁ClN₂O₃
MW: 182.604 g/mol
MDL Number: MFCD00039056
CAS Number: 2776-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty