Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di(naphthalen-2-yl)phosphine oxide 97%

Supplier: AMBEED, INC
Warning

A109288-1G A109288-250MG A109288-25G A109288-5G
77093-882EA 40 USD InStock
77093-882 77093-884 77093-886 77093-888
Di(naphthalen-2-yl)phosphine oxide 97%
Di(naphthalen-2-yl)phosphine oxide
Precaución: for Research Use Only


Formula: C₂₀H₁₅OP
MW: 302.31 g/mol
MDL Number: MFCD29477760
CAS Number: 78871-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty