Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(4-Methylpiperazin-1-yl)-4-(o-tolyl)nicotinamide 95%

Supplier: AMBEED, INC
Warning

A110317-100MG A110317-1G A110317-250MG A110317-5G
77093-976EA 5 USD InStock
77093-976 77093-978 77093-980 77093-982
6-(4-Methylpiperazin-1-yl)-4-(o-tolyl)nicotinamide 95%
6-(4-Methylpiperazin-1-yl)-4-(o-tolyl)nicotinamide
Precaución: for Research Use Only


Formula: C₁₈H₂₂N₄O
MDL Number: MFCD11113419
CAS Number: 342417-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty