Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ORY-1001 98%

Supplier: AMBEED, INC
Warning

A114267-10MG A114267-1MG A114267-2MG A114267-5MG A114267-100MG A114267-50MG
77094-132EA 95 USD InStock
77094-132 77094-134 77094-136 77094-138 77685-011 77685-012
ORY-1001 98%
ORY-1001
Precaución: for Research Use Only


Formula: C₁₅H₂₄Cl₂N₂
MDL Number: MFCD28900684
CAS Number: 1431326-61-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty