Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-amino-4-bromo-2,6-difluorobenzoate 95+%

Supplier: AMBEED, INC
Warning

A118010-1G A118010-250MG A118010-5G
77094-308EA 40 USD InStock
77094-308 77094-310 77094-312
Methyl 3-amino-4-bromo-2,6-difluorobenzoate 95+%
Methyl 3-amino-4-bromo-2,6-difluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₆BrF₂NO₂
MW: 266.03 g/mol
MDL Number: MFCD23840403
CAS Number: 1529613-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty