Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-chloropyridine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A118117-10G A118117-25G A118117-5G
77094-324EA 18 USD InStock
77094-324 77094-326 77094-328
3-Bromo-4-chloropyridine hydrochloride 97%
3-Bromo-4-chloropyridine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₄BrCl₂N
MW: 228.91 g/mol
MDL Number: MFCD08448229
CAS Number: 181256-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty