Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Cysteine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A120586-100G A120586-5G
77094-430EA 50 USD InStock
77094-430 77094-432
D-Cysteine hydrochloride 98%
D-Cysteine hydrochloride
Precaución: for Research Use Only


Formula: C₃H₇NO₂S·HCl
MDL Number: MFCD00012632
CAS Number: 32443-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty