Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-formylsalicylate 98%

Supplier: AMBEED, INC
Warning

A121024-100G A121024-1G A121024-25G A121024-5G
77094-444EA 187 USD InStock
77094-444 77094-446 77094-448 77094-450
Methyl-5-formylsalicylate 98%
Methyl 5-formylsalicylate
Precaución: for Research Use Only


Formula: C₉H₈O₄
MDL Number: MFCD03840784
CAS Number: 41489-76-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty