Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-((phenylamino)methylene)malonate 95%

Supplier: AMBEED, INC
Warning

A121458-10G A121458-1G A121458-250MG A121458-25G A121458-5G
77094-478EA 178 USD InStock
77094-478 77094-480 77094-482 77094-484 77094-486
Diethyl 2-((phenylamino)methylene)malonate 95%
Diethyl 2-((phenylamino)methylene)malonate
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₄
MW: 263.29 g/mol
MDL Number: MFCD00173404
CAS Number: 54535-22-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty