Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoropicolinic acid 98%

Supplier: AMBEED, INC
Warning

A121858-10G A121858-1G A121858-25G A121858-5G A121858-100G
77094-504EA 32 USD InStock
77094-504 77094-506 77094-508 77094-510 77685-076
3-Fluoropicolinic acid 98%
3-Fluoropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₄FNO₂
MW: 141.1
MDL Number: MFCD04114108
CAS Number: 152126-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty