Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Allyloxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A123970-100MG A123970-1G A123970-250MG
77094-640EA 14 USD InStock
77094-640 77094-642 77094-644
3-Allyloxyphenylboronic acid 97%
3-Allyloxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BO₃
MDL Number: MFCD11103733
CAS Number: 222840-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty