Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminodecanoic acid 95%

Supplier: AMBEED, INC
Warning

A124957-100MG A124957-250MG
77094-698EA 46 USD InStock
77094-698 77094-700
2-Aminodecanoic acid 95%
2-Aminodecanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₂₁NO₂
MW: 187.28 g/mol
MDL Number: MFCD01862887
CAS Number: 17702-88-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty