Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethyl-4-piperidone 97%

Supplier: AMBEED, INC
Warning

A126350-100G A126350-10G A126350-25G A126350-5G
77094-796EA 73 USD InStock
77094-796 77094-798 77094-800 77685-102
1,3-Dimethyl-4-piperidone 97%
1,3-Dimethyl-4-piperidone
Precaución: for Research Use Only


Formula: C₇H₁₃NO
MDL Number: MFCD00100893
CAS Number: 4629-80-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty