Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidin-2-ol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A127216-100MG A127216-250MG
77094-822EA 150 USD InStock
77094-822 77094-824
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidin-2-ol hydrochloride 95%
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidin-2-ol hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉N₃O·ClH
MDL Number: MFCD16615181
CAS Number: 1956321-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty