Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dichloro-2-methylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A127351-100MG A127351-1G A127351-250MG
77094-826EA 10 USD InStock
77094-826 77094-828 77094-830
4,5-Dichloro-2-methylbenzoic acid 97%
4,5-Dichloro-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O₂
MDL Number: MFCD22566295
CAS Number: 5252-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty