Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Adamantan-1-yl)-4-methyl-1H-pyrrole 97%

Supplier: AMBEED, INC
Warning

A133320-1G A133320-250MG A133320-5G
77095-052EA 37.14 USD InStock
77095-052 77095-054 77095-056 77095-058
2-(Adamantan-1-yl)-4-methyl-1H-pyrrole 97%
2-(Adamantan-1-yl)-4-methyl-1H-pyrrole
Precaución: for Research Use Only


Formula: C₁₅H₂₁N
MDL Number: MFCD29477676
CAS Number: 1914148-52-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty