Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9,10-Di(1-naphthyl)anthracene 97%

Supplier: AMBEED, INC
Warning

A137615-1G A137615-250MG A137615-25G A137615-5G
77095-276EA 21 USD InStock
77095-276 77095-278 77095-280 77095-282
9,10-Di(1-naphthyl)anthracene 97%
9,10-Di(1-naphthyl)anthracene
Precaución: for Research Use Only


Formula: C₃₄H₂₂
MDL Number: MFCD04222155
CAS Number: 26979-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty