Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methyl-5-nitroaniline 97%

Supplier: AMBEED, INC
Warning

A137930-100MG A137930-1G A137930-250MG
77095-292EA 6 USD InStock
77095-292 77095-294 77095-296
4-Fluoro-2-methyl-5-nitroaniline 97%
4-Fluoro-2-methyl-5-nitroaniline
Precaución: For research use only.


Formula: C₇H₇FN₂O₂
MDL Number: MFCD18824947
CAS Number: 446-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty