Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2'-Amino-1,1'-biphenyl-2-yl)methanesulfonatopalladium(II) dimer 98%

Supplier: AMBEED, INC
Warning

A139122-100MG A139122-1G A139122-250MG
77095-366EA 70 USD InStock
77095-366 77095-368 77095-370
(2'-Amino-1,1'-biphenyl-2-yl)methanesulfonatopalladium(II) dimer 98%
(2'-Amino-1,1'-biphenyl-2-yl)methanesulfonatopalladium(II) dimer
Precaución: for Research Use Only


Formula: C₂₆H₂₆N₂O₆Pd₂S₂
MDL Number: MFCD22666044
CAS Number: 1435520-65-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty