Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acalabrutinib 98%

Supplier: AMBEED, INC
Warning

A141995-1G A141995-1MG A141995-2MG A141995-5MG A141995-100MG
77095-460EA 58 USD InStock
77095-460 77095-462 77095-464 77095-466 77685-210
Acalabrutinib 98%
Acalabrutinib
Precaución: for Research Use Only


Formula: C₂₆H₂₃N₇O₂
MDL Number: MFCD29472294
CAS Number: 1420477-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty