Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrogallol red 98%

Supplier: AMBEED, INC
Warning

A155745-1G A155745-250MG A155745-5G
77095-962EA 45 USD InStock
77095-962 77095-964 77095-966
Pyrogallol red 98%
Pyrogallol red
Precaución: for Research Use Only


Formula: C₁₉H₁₂O₈S
MW: 400.36 g/mol
MDL Number: MFCD00005046
CAS Number: 32638-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty