Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluorobenzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A155827-1G A155827-250MG A155827-5G
77095-972EA 11 USD InStock
77095-972 77095-974 77095-976
3,4-Difluorobenzenesulphonamide 98%
3,4-Difluorobenzenesulphonamide
Precaución: for Research Use Only


Formula: C₆H₅F₂NO₂S
MW: 193.17
MDL Number: MFCD00728800
CAS Number: 108966-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty