Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Benzyl-D-glutamate 98%

Supplier: AMBEED, INC
Warning

A158912-100G A158912-10G A158912-1G A158912-25G A158912-500G A158912-5G
77096-090EA 103 USD InStock
77096-090 77096-092 77096-094 77096-096 77096-098 77096-100
5-Benzyl-D-glutamate 98%
5-Benzyl-D-glutamate
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00063111
CAS Number: 2578-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty