Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorophenol 98%

Supplier: AMBEED, INC
Danger

A159857-10G A159857-25G A159857-5G
77096-110EA 19 USD InStock
77096-110 77096-112 77096-114
2,6-Difluorophenol 98%
2,6-Difluorophenol
Precaución: for Research Use Only


Formula: C₆H₄F₂O
MDL Number: MFCD00002158
CAS Number: 28177-48-2
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty