Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-((S)-3,3-Dimethyl-1-(methylamino)-1-oxobutan-2-yl)-2-((S)-1-(N-hydroxyformamido)ethyl)-5-phenylpentanamide 95%

Supplier: AMBEED, INC
Warning

A164330-1MG A164330-5MG
77096-252EA 55 USD InStock
77096-252 77096-254
(R)-N-((S)-3,3-Dimethyl-1-(methylamino)-1-oxobutan-2-yl)-2-((S)-1-(N-hydroxyformamido)ethyl)-5-phenylpentanamide 95%
(R)-N-((S)-3,3-Dimethyl-1-(methylamino)-1-oxobutan-2-yl)-2-((S)-1-(N-hydroxyformamido)ethyl)-5-phenylpentanamide
Precaución: for Research Use Only


Formula: C₂₁H₃₃N₃O₄
MDL Number: MFCD19381832
CAS Number: 260264-93-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty