Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromobicyclo[4.2.0]octa-1,3,5-triene 98%

Supplier: AMBEED, INC
Warning

A164818-100MG A164818-250MG A164818-25G A164818-5G
77096-256EA 5 USD InStock
77096-256 77096-258 77096-260 77096-262
7-Bromobicyclo[4.2.0]octa-1,3,5-triene 98%
7-Bromobicyclo[4.2.0]octa-1,3,5-triene
Precaución: for Research Use Only


Formula: C₈H₇Br
MDL Number: MFCD17976891
CAS Number: 21120-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty