Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Formyl-4-picoline 98%

Supplier: AMBEED, INC
Warning

A166216-100G A166216-1G A166216-250MG A166216-25G A166216-5G
77096-322EA 481 USD InStock
77096-322 77096-324 77096-326 77096-328 77096-330
2-Formyl-4-picoline 98%
2-Formyl-4-picoline
Precaución: for Research Use Only


Formula: C₇H₇NO
MW: 121.1366 g/mol
MDL Number: MFCD07367933
CAS Number: 53547-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty