Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloroaniline 98%

Supplier: AMBEED, INC
Warning

A167628-100G A167628-10G A167628-25G A167628-500G A167628-5G
77096-398EA 42 USD InStock
77096-398 77096-400 77096-402 77096-404 77096-406
4-Bromo-2-chloroaniline 98%
4-Bromo-2-chloroaniline
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MDL Number: MFCD00007660
CAS Number: 38762-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty