Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GDC0994 ≥98%

Supplier: AMBEED, INC
Warning

A170354-10MG A170354-1MG A170354-2MG A170354-50MG A170354-5MG
77096-484EA 90 USD InStock
77096-484 77096-486 77096-488 77096-490 77096-492
GDC0994 ≥98%
GDC0994
Precaución: for Research Use Only


Formula: C₂₁H₁₈ClFN₆O₂
MDL Number: MFCD28143918
CAS Number: 1453848-26-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty