Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CTx-0294885 98%

Supplier: AMBEED, INC
Warning

A173517-100MG A173517-10MG A173517-1MG A173517-250MG A173517-2MG A173517-5MG A173517-50MG
77096-558EA 241 USD InStock
77096-558 77096-560 77096-562 77096-564 77096-566 77096-568 77685-388
CTx-0294885 98%
CTx-0294885
Precaución: for Research Use Only


Formula: C₂₂H₂₄ClN₇O
MW: 437.93 g/mol
MDL Number: MFCD28137930
CAS Number: 1439934-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty