Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-alanine benzyl ester 95%

Supplier: AMBEED, INC
Warning

A177772-10G A177772-1G A177772-25G A177772-5G
77096-704EA 21 USD InStock
77096-704 77096-706 77096-708 77096-710
Boc-L-alanine benzyl ester 95%
Boc-L-alanine benzyl ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₄
MDL Number: MFCD00190775
CAS Number: 51814-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty