Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PF-05212384 ≥98%

Supplier: AMBEED, INC
Warning

A178133-10MG A178133-1MG A178133-5MG
77096-742EA 54 USD InStock
77096-742 77096-744 77096-746
PF-05212384 ≥98%
PF-05212384
Precaución: for Research Use Only


Formula: C₃₂H₄₁N₉O₄
MW: 615.73 g/mol
MDL Number: MFCD16875679
CAS Number: 1197160-78-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty