Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl dodecanedioate 98%

Supplier: AMBEED, INC
Warning

A180076-100G A180076-25G A180076-5G A180076-500G A180076-1000G
77096-798EA 22 USD InStock
77096-798 77096-800 77096-802 77685-412 77685-411
Dimethyl dodecanedioate 98%
Dimethyl dodecanedioate
Precaución: for Research Use Only


Formula: C₁₄H₂₆O₄
MDL Number: MFCD00043632
CAS Number: 1731-79-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty