Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-iodoaniline 98%

Supplier: AMBEED, INC
Warning

A181026-100G A181026-10G A181026-25G A181026-500G
77096-840EA 42 USD InStock
77096-840 77096-842 77096-844 77096-846
2-Fluoro-4-iodoaniline 98%
2-Fluoro-4-iodoaniline
Precaución: for Research Use Only


Formula: C₆H₅FIN
MDL Number: MFCD00011738
CAS Number: 29632-74-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty