Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoquinoline-7-amine 98%

Supplier: AMBEED, INC
Warning

A181329-100MG A181329-10G A181329-1G A181329-250MG
77096-862EA 10 USD InStock
77096-862 77096-864 77096-866 77096-868
Isoquinoline-7-amine 98%
Isoquinoline-7-amine
Precaución: for Research Use Only


Formula: C₉H₈N₂
MDL Number: MFCD08669932
CAS Number: 23707-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty