Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pent-4-yn-1-amine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A184504-1G A184504-250MG A184504-5G
77096-960EA 111 USD InStock
77096-960 77096-962 77685-431
Pent-4-yn-1-amine hydrochloride 95%
Pent-4-yn-1-amine hydrochloride
Precaución: For research use only.


Formula: C₅H₉N·ClH
MDL Number: MFCD22836507
CAS Number: 173987-24-1
UN: 3263
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty