Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane 97%

Supplier: AMBEED, INC
Warning

A186693-10G A186693-1G A186693-25G A186693-5G
77097-030EA 63 USD InStock
77097-030 77097-032 77097-034 77097-036
9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane 97%
9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO
MDL Number: MFCD00144851
CAS Number: 2291-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty