Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-hydroxy-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 95%

Supplier: AMBEED, INC
Warning

A187068-1G A187068-250MG
77097-038EA 86 USD InStock
77097-038 77097-040
tert-Butyl 4-hydroxy-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 95%
tert-Butyl 4-hydroxy-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD11869803
CAS Number: 1142188-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty