Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98%, ee 99%

Supplier: AMBEED, INC
Danger

A187955-100MG A187955-1G A187955-250MG
77097-064EA 31 USD InStock
77097-064 77097-066 77685-444
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98%, ee 99%
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine
Precaución: for Research Use Only


Formula: C₁₆H₁₆N₂
MDL Number: MFCD09038520
CAS Number: 181139-49-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty