Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanophenylacetonitrile 98%

Supplier: AMBEED, INC
Warning

A189520-100G A189520-10G A189520-25G A189520-5G A189520-1G
77097-090EA 117 USD InStock
77097-090 77097-092 77097-094 77097-096 77685-450
2-Cyanophenylacetonitrile 98%
2-Cyanophenylacetonitrile
Precaución: for Research Use Only


Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00001895
CAS Number: 3759-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty