Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-1,4-dimethoxybenzene 95%

Supplier: AMBEED, INC
Warning

A195330-100MG A195330-250MG
77097-220EA 10 USD InStock
77097-220 77097-222
2-Iodo-1,4-dimethoxybenzene 95%
2-Iodo-1,4-dimethoxybenzene
Precaución: for Research Use Only


Formula: C₈H₉IO₂
MDL Number: MFCD09032417
CAS Number: 25245-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty