Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-((tert-butoxycarbonyl)amino)-2-methylpropanoate 95%

Supplier: AMBEED, INC
Warning

A197647-100MG A197647-1G A197647-250MG A197647-25G A197647-5G
77097-294EA 5 USD InStock
77097-294 77097-296 77097-298 77097-300 77097-302
Methyl 2-((tert-butoxycarbonyl)amino)-2-methylpropanoate 95%
Methyl 2-((tert-butoxycarbonyl)amino)-2-methylpropanoate
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₄
MDL Number: MFCD20484740
CAS Number: 84758-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty