Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Isoleucinol 97%

Supplier: AMBEED, INC
Warning

A199688-100G A199688-1G A199688-25G A199688-500G
77097-376EA 55 USD InStock
77097-376 77097-378 77097-380 77097-382
L(+)-Isoleucinol 97%
L(+)-Isoleucinol
Precaución: For research use only.


Formula: C₆H₁₅NO
MDL Number: MFCD00004731
CAS Number: 24629-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty