Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1-amino-2,3-dihydro-1H-indene-4-carboxylate 98%

Supplier: AMBEED, INC
Warning

A211725-100MG A211725-1G A211725-250MG
77097-694EA 151 USD InStock
77097-694 77097-696 77097-698
Methyl 1-amino-2,3-dihydro-1H-indene-4-carboxylate 98%
Methyl 1-amino-2,3-dihydro-1H-indene-4-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₂
MW: 191.22 g/mol
MDL Number: MFCD18650505
CAS Number: 1273664-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty