Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-nitroaniline 97%

Supplier: AMBEED, INC
Danger

A212838-100G A212838-10G A212838-1G A212838-25G A212838-5G
77097-724EA 183 USD InStock
77097-724 77097-726 77097-728 77097-730 77097-732
3-Chloro-4-nitroaniline 97%
3-Chloro-4-nitroaniline
Precaución: for Research Use Only


Formula: C6H5ClN2O2
MW: 172.57 g/mol
MDL Number: MFCD00085922
CAS Number: 825-41-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty