Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CHS-828 98%

Supplier: AMBEED, INC
Warning

A215361-10MG A215361-1MG A215361-5MG
77097-804EA 44 USD InStock
77097-804 77097-806 77097-808
CHS-828 98%
CHS-828
Precaución: for Research Use Only


Formula: C₁₉H₂₂ClN₅O
MDL Number: MFCD03700766
CAS Number: 200484-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty